α-Chloro-3,4-difluorotoulene
Supplier: Matrix Scientific
Synonyms:
1-(Chloromethyl)-3,4-difluorobenzene, 3,4-Difluorobenzyl chloride, 4-(Chloromethyl)-1,2-difluorobenzene
005241-5G
005241-25G
101778-142EA
23.08
USD
101778-142
101778-144
α-Chloro-3,4-difluorotoulene
α-Chloro-3,4-difluorotoulene
Hazard: Corrosive/Lachrymator
Formula:
C₇H₅ClF₂ MW: 162.57 g/mol Boiling Pt: 28°C/ 0.4mm |
MDL Number:
MFCD00236218 CAS Number: 698-80-6 UN: 1760 ADR: 8,III |
Specification Test Results
Boiling point | 28°/ 0.4mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...