α,3,5-Trichlorotoluene ≥98%

Supplier: Matrix Scientific

005240-25G 005240-5G 005240-1G
101778-140EA 253.85 USD
101778-140 101778-138 101778-136
α,3,5-Trichlorotoluene ≥98%
α,3,5-Trichlorotoluene

Hazard: Corrosive/Lachrymator

Formula: C₇H₅Cl₃
Melting Pt: 32°C
MDL Number: MFCD00060278
CAS Number: 3290-06-0

Order Now

Specification Test Results

Melting point 32°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR