α-Bromo-2,6-difluorotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Bromomethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl bromide

005233-5G 005233-25G
101778-120EA 20 USD
101778-120 101778-122
α-Bromo-2,6-difluorotoluene ≥97%
α-Bromo-2,6-difluorotoluene

Hazard: Corrosive/Lachrymator

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Melting Pt: 53…55 °C
MDL Number: MFCD00000329
CAS Number: 85118-00-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 53-55°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR