2,5-Difluorobenzoyl chloride ≥96%

Supplier: Matrix Scientific

005230-25G 005230-100G
101778-116EA 67.69 USD
101778-116 101778-118
2,5-Difluorobenzoyl chloride ≥96%
2,5-Difluorobenzoyl chloride

Hazard: Corrosive/Lachrymator

Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Boiling Pt: 92…93 °C (34 mmHg)
Density: 1.425
MDL Number: MFCD00009929
CAS Number: 35730-09-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 92-93°/34m
Density 1.425
ND20 1.515

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR