2,6-Difluorobenzoyl chloride ≥98%
Supplier: Matrix Scientific
005229-100G
005229-25G
101778-114EA
280
USD
101778-114
101778-112
2,6-Difluorobenzoyl chloride ≥98%
2,6-Difluorobenzoyl chloride
Hazard: Corrosive/Lachrymator
Formula:
C₇H₃ClF₂O MW: 176.55 g/mol Boiling Pt: 72…75 °C (13 mmHg) Density: 1.404 |
MDL Number:
MFCD00000659 CAS Number: 18063-02-0 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 72-75°/13m |
Density | 1.404 |
ND20 | 1.499 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...