2,4-Difluorobenzoyl chloride ≥98%

Supplier: Matrix Scientific

005228-100G 005228-25G
101778-110EA 209.23 USD
101778-110 101778-108
2,4-Difluorobenzoyl chloride ≥98%
2,4-Difluorobenzoyl chloride

Hazard: Corrosive/Lachrymator

Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Boiling Pt: 192°C
Density: 1.437
MDL Number: MFCD00000658
CAS Number: 72482-64-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 192°
Density 1.437
ND20 1.516

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR