3,4-Difluorobenzoyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3,4-difluorobenzoylchloride

005226-100G 005226-25G
101778-106EA 269.23 USD
101778-106 101778-104
3,4-Difluorobenzoyl chloride ≥97%
3,4-Difluorobenzoyl chloride

Hazard: Corrosive/Lachrymator

Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Boiling Pt: 186 °C
Density: 1.403
MDL Number: MFCD00009930
CAS Number: 76903-88-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 186°
Density 1.403
ND20 1.5114

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR