3,5-Difluorobenzoyl chloride ≥98%
Supplier: Matrix Scientific
005224-25G
005224-100G
101778-100EA
53.85
USD
101778-100
101778-102
3,5-Difluorobenzoyl chloride ≥98%
3,5-Difluorobenzoyl chloride
Hazard: Corrosive/Stored under Argon/Lachrymator
Formula:
C₇H₃ClF₂O MW: 176.55 g/mol |
MDL Number:
MFCD00010309 CAS Number: 129714-97-2 UN: 3265 ADR: 8,II |
Specification Test Results
ND20 | 1.5031 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...