3,4-Dichloro-α,α,α-trifluorotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 1,2-Dichloro-4-(trifluoromethyl)benzene, 3,4-Dichlorobenzotrifluoride

005223-250G 005223-100G
101778-098EA 72.31 USD
101778-098 101778-096
3,4-Dichloro-α,α,α-trifluorotoluene ≥97%
3,4-Dichloro-α,α,α-trifluorotoluene

Hazard: Corrosive

Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
Boiling Pt: 173-175°C
Density: 1.478
MDL Number: MFCD00000555
CAS Number: 328-84-7
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 173-175°
Density 1.478
ND20 1.475

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR