α,α,α,3,4-Pentafluorotoluene

Supplier: Matrix Scientific

005220-1G 005220-5G 005220-25G
101778-090EA 12.31 USD
101778-090 101778-092 101778-094
α,α,α,3,4-Pentafluorotoluene
α,α,α,3,4-Pentafluorotoluene

Hazard: Flammable

Formula: C₇H₃F₅
Boiling Pt: 104°C
Melting Pt: 95-98°C
MDL Number: MFCD00077510
CAS Number: 32137-19-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR