3,4-Difluorobenzophenone ≥98%

Supplier: Matrix Scientific

005210-5G 005210-25G
101778-082EA 46.15 USD
101778-082 101778-084
3,4-Difluorobenzophenone ≥98%
3,4-Difluorobenzophenone

Hazard: Irritant

Formula: C₁₃H₈F₂O
Melting Pt: 53-55°C
MDL Number: MFCD00009892
CAS Number: 85118-07-6

Order Now

Specification Test Results

Melting point 53-55°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR