2,3-Dichlorobenzenesulfonyl chloride ≥98%
Supplier: Matrix Scientific
005184-5G
005184-25G
101778-066EA
23.08
USD
101778-066
101778-068
2,3-Dichlorobenzenesulfonyl chloride ≥98%
2,3-Dichlorobenzenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₆H₃Cl₃O₂S Boiling Pt: 140…142 °C (2 mmHg) Melting Pt: 61…63 °C |
MDL Number:
MFCD00051844 CAS Number: 82417-45-6 UN: 3261 ADR: 8,II |
Specification Test Results
Boiling point | 140-142°/2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...