3,4-Difluorobenzenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
Synonyms:
3,4-Difluorobenzene-1-sulphonyl chloride
005176-5G
005176-25G
101778-050EA
20
USD
101778-050
101778-052
3,4-Difluorobenzenesulfonyl chloride ≥97%
3,4-Difluorobenzenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₆H₃ClF₂O₂S MW: 212.6 g/mol Boiling Pt: 105°C/15 Torr Density: 1.586 |
MDL Number:
MFCD00153120 CAS Number: 145758-05-0 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...