2,6-Dichlorobenzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

005174-25G 005174-5G
101778-044EA 80 USD
101778-044 101778-042
2,6-Dichlorobenzenesulfonyl chloride ≥95%
2,6-Dichlorobenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₃Cl₃O₂S
Melting Pt: 52…54 °C
MDL Number: MFCD00052311
CAS Number: 6579-54-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 52-54°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR