2,5-Dichlorobenzenesulphonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2,5-Dichlorobenzenesulphonyl Chloride, 2,5-Dichlorobenzene-1-sulphonyl chloride

005173-25G 005173-100G
101778-038EA 30.77 USD
101778-038 101778-040
2,5-Dichlorobenzenesulphonyl chloride ≥98%
2,5-Dichlorobenzenesulphonyl chloride

TCA listed: T
Hazard: Corrosive/Stored under Argon/Moisture Sensitive

Formula: C₆H₃Cl₃O₂S
Melting Pt: 36-37°C
MDL Number: MFCD00007429
CAS Number: 5402-73-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 36-37°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR