2,4-Difluorobenzaldehyde ≥98%

Supplier: Matrix Scientific

005163-100G 005163-25G
101778-032EA 258.46 USD
101778-032 101778-030
2,4-Difluorobenzaldehyde ≥98%
2,4-Difluorobenzaldehyde

Hazard: Flammable/Irritant

Formula: C₇H₄F₂O
Boiling Pt: 65…66 °C (17 mmHg)
Melting Pt: 2…3 °C
Density: 1.299
MDL Number: MFCD00010326
CAS Number: 1550-35-2
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Boiling point 65-66°/17m
Density 1.299
ND20 1.498


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR