2,4-Difluoroanisole ≥99%

Supplier: Matrix Scientific

Synonyms: 2,4-Difluoro-1-methoxybenzene

005157-25G 005157-100G
101778-026EA 29.23 USD
101778-026 101778-028
2,4-Difluoroanisole ≥99%
2,4-Difluoroanisole

Hazard: Flammable

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 53 °C (18 mmHg)
Density: 1.246
MDL Number: MFCD00010731
CAS Number: 452-10-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 53°/18mm
Density 1.246
ND20 1.472

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR