2,6-Difluoroanisole ≥98%

Supplier: Matrix Scientific

005155-5G 005155-25G
101778-022EA 52.31 USD
101778-022 101778-024
2,6-Difluoroanisole ≥98%
2,6-Difluoroanisole

Hazard: Flammable

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 62 °C (40 Torr)
Density: 1.221
MDL Number: MFCD00142846
CAS Number: 437-82-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR