2,3-Difluoroanisole ≥99%
Supplier: Matrix Scientific
005154-1G
005154-5G
005154-25G
101778-016EA
13.85
USD
101778-016
101778-018
101778-020
2,3-Difluoroanisole ≥99%
2,3-Difluoroanisole
Hazard: Flammable
Formula:
C₇H₆F₂O MW: 144.12 g/mol Boiling Pt: 70…72 °C (38 mmHg) |
MDL Number:
MFCD01631562 CAS Number: 134364-69-5 |
Specification Test Results
Boiling point | 70-72°/38 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...