3,5-Difluoroanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 1,3-Difluoro-5-methoxybenzene

005152-5G 005152-25G
101778-008EA 27.69 USD
101778-008 101778-010
3,5-Difluoroanisole ≥98%
3,5-Difluoroanisole

Hazard: Flammable

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 145-148°C
Density: 1.234
Flash Pt: 43°C
MDL Number: MFCD00042560
CAS Number: 93343-10-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Density 1.234
ND20 1.466

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR