2,5-Difluoroanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 1,4-Difluoro-2-methoxybenzene

005151-5G 005151-25G
101778-004EA 653.85 USD
101778-004 101778-006
2,5-Difluoroanisole ≥98%
2,5-Difluoroanisole

Hazard: Flammable

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 72-73°C/25mm
Flash Pt: 40°C
MDL Number: MFCD00042561
CAS Number: 75626-17-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR