3,4-Difluoroaniline ≥99%
Supplier: Matrix Scientific
005150-100G
005150-500G
101778-000EA
83.08
USD
101778-000
101778-002
3,4-Difluoroaniline ≥99%
3,4-Difluoroaniline
Hazard: Toxic/Irritant/Refrigerate
Formula:
C₆H₅F₂N Boiling Pt: 77°C/7 Torr Melting Pt: 227-228°C Density: 1.302 |
MDL Number:
MFCD00007761 CAS Number: 3863-11-4 UN: 2941 ADR: 6.1,III |
Specification Test Results
Boiling point | 77°/7mm |
Density | 1.302 |
ND20 | 1.513 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...