2,4-Difluoroaniline ≥99%
Supplier: Matrix Scientific
005146-100G
005146-500G
101777-984EA
20
USD
101777-984
101777-986
2,4-Difluoroaniline ≥99%
2,4-Difluoroaniline
TCA listed: T
Hazard: Toxic/Irritant
Formula:
C₆H₅F₂N MW: 129.11 g/mol Boiling Pt: 168…170 °C Melting Pt: –7.5 °C Density: 1.268 |
MDL Number:
MFCD00007648 CAS Number: 367-25-9 UN: 2941 ADR: 6.1,III |
Specification Test Results
Boiling point | 168-170° |
Density | 1.268 |
ND20 | 1.507 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...