2,2-Difluoroacetyl chloride
Supplier: Matrix Scientific
005140-25G
005140-100G
101777-970EA
861.54
USD
101777-970
101777-972
2,2-Difluoroacetyl chloride
2,2-Difluoroacetyl chloride
Hazard: Corrosive/Store in Freezer
Formula:
C₂HClF₂O Boiling Pt: 25°C Density: |
CAS Number:
381-72-6 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 25° |
Density |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...