3,4-Dichloro-5-nitrobenzotrifluoride ≥97%

Supplier: Matrix Scientific

005120-1G 005120-5G
101777-952EA 60 USD
101777-952 101777-954
3,4-Dichloro-5-nitrobenzotrifluoride ≥97%
3,4-Dichloro-5-nitrobenzotrifluoride

Hazard: Irritant/Irritant-Harmful

Formula: C₇H₂Cl₂F₃NO₂
Boiling Pt: 115…118 °C (15 Torr)
MDL Number: MFCD01862035
CAS Number: 657-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR