2,4-Dichloro-5-methylpyrimidine ≥98%

Supplier: Matrix Scientific

005117-25G 005117-100G
101777-950EA 33.85 USD
101777-950 102832-762
2,4-Dichloro-5-methylpyrimidine ≥98%
2,4-Dichloro-5-methylpyrimidine

TCA listed: T
Hazard: Corrosive/Avoid Skin Contact/Lachrymator

Formula: C₅H₄Cl₂N₂
Boiling Pt: 108…109 °C (11 mmHg)
Melting Pt: 26…28 °C
Density: 1.39
MDL Number: MFCD00023197
CAS Number: 1780-31-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 108-109°/1
Density 1.39


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR