2,4-Dichloro-5-fluorobenzoyl chloride
Supplier: Matrix Scientific
005111-5G
005111-25G
101777-944EA
44.62
USD
101777-944
101777-946
2,4-Dichloro-5-fluorobenzoyl chloride
2,4-Dichloro-5-fluorobenzoyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₇H₂Cl₃FO Boiling Pt: 143…144 °C (35 mmHg) Density: 1.568 |
MDL Number:
MFCD00075341 CAS Number: 86393-34-2 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 143-144°/3 |
Density | 1.568 |
ND20 | 1.572 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...