2,3-Difluoro-4-methylbenzyl bromide

Supplier: Matrix Scientific

005077-1G 005077-5G
101777-932EA 60 USD
101777-932 101777-934
2,3-Difluoro-4-methylbenzyl bromide
2,3-Difluoro-4-methylbenzyl bromide

Hazard: Corrosive

Formula: C₈H₇BrF₂
MW: 221.04 g/mol
Boiling Pt: 78°C/1 Torr
MDL Number: MFCD01631647
CAS Number: 261763-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR