2,6-Dichloro-4-(trifluoromethoxy)aniline
Supplier: Matrix Scientific
005032-5G
005032-25G
101777-912EA
43.08
USD
101777-912
101777-914
2,6-Dichloro-4-(trifluoromethoxy)aniline
2,6-Dichloro-4-(trifluoromethoxy)aniline
Hazard: Toxic
Formula:
C₇H₄Cl₂F₃NO Melting Pt: 30-32°C |
MDL Number:
MFCD00190128 CAS Number: 99479-66-0 |
Specification Test Results
Melting point | 30-32° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...