2,6-Difluoro-3-toluoyl chloride

Supplier: Matrix Scientific

005016-1G 005016-5G
101777-900EA 30.77 USD
101777-900 101777-902
2,6-Difluoro-3-toluoyl chloride
2,6-Difluoro-3-toluoyl chloride

Hazard: Corrosive

Formula: C₈H₅ClF₂O
MDL Number: MFCD01631334
CAS Number: 261763-39-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR