1,3-Difluoro-2-propanol stabilized
Supplier: Matrix Scientific
Stabilization:
Stabilised with sodium fluoride
005001-1G
005001-5G
101777-894EA
23.08
USD
101777-894
101777-896
1,3-Difluoro-2-propanol stabilized
1,3-Difluoro-2-propanol
TCA listed: T
Hazard: Flammable/Toxic
Formula:
C₃H₆F₂O MW: 96.08 g/mol Boiling Pt: 127 °C Density: 1.244 Flash Pt: 42 °C |
MDL Number:
MFCD00000452 CAS Number: 453-13-4 UN: 1986 ADR: 3,III |
Specification Test Results
Boiling point | 127° |
Density | 1.244 |
ND20 | 1.373 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...