DFDNB (1,5-Difluoro-2,4-dinitrobenzene) ≥97%

Supplier: Matrix Scientific

004983-100G 004983-25G
101777-892EA 869.23 USD
101777-892 101777-890
DFDNB (1,5-Difluoro-2,4-dinitrobenzene) ≥97%
DFDNB (1,5-Difluoro-2,4-dinitrobenzene)

TCA listed: T
Hazard: Toxic

Formula: C₆H₂F₂N₂O₄
Melting Pt: 74…75 °C
MDL Number: MFCD00007052
CAS Number: 327-92-4
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Melting point 74-75°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR