3,5-Dichloro-2,4,6-trifluoropyridine ≥98%
Supplier: Matrix Scientific
Synonyms:
3,5-Dichlorotrifluoropyridine
004982-25G
004982-100G
101777-886EA
49.23
USD
101777-886
101777-888
3,5-Dichloro-2,4,6-trifluoropyridine ≥98%
3,5-Dichloro-2,4,6-trifluoropyridine
TCA listed: T
Hazard: Toxic by inhalation
Formula:
C₅Cl₂F₃N MW: 201.96 g/mol Boiling Pt: 159…160 °C Melting Pt: 24…26 °C Density: 1.622 |
MDL Number:
MFCD00006226 CAS Number: 1737-93-5 UN: 2811 ADR: 6.1,II |
Specification Test Results
Boiling point | 159-160° |
Density | 1.622 |
ND20 | 1.478 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...