1,1'-Carbonyldiimidazole ≥98%
Supplier: Matrix Scientific
Synonyms:
Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI
004914-250G
004914-1KG
101777-846EA
146.15
USD
101777-846
101777-848
1,1'-Carbonyldiimidazole ≥98%
1,1'-Carbonyldiimidazole
TCA listed: T
Hazard: Corrosive/Stored under Argon/Moisture Sensitive/Refrigerate
Formula:
C₇H₆N₄O MW: 162.15 g/mol Melting Pt: 117…122 °C |
MDL Number:
MFCD00005286 CAS Number: 530-62-1 UN: 3263 ADR: 8,II |
Specification Test Results
Melting point | 117-122° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...