Bromoacetonitrile ≥98%

Supplier: Matrix Scientific

004912-25G 004912-100G
101777-842EA 58.46 USD
101777-842 101777-844
Bromoacetonitrile ≥98%
Bromoacetonitrile

Hazard: Toxic/Corrosive/Refrigerate/Lachrymator

Formula: BrCNCH₂
Boiling Pt: 60…62 °C (24 mmHg)
Density: 1.722
MDL Number: MFCD00001884
CAS Number: 590-17-0
UN: 3276
ADR: 6.1,II

Order Now

Specification Test Results

Boiling point 60-62°/24m
Density 1.722
ND20 1.48


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR