4-Fluorotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Fluoro-4-methylbenzene

004893-500G 004893-100G 004893-2.5KG
101777-830EA 36.92 USD
101777-830 101777-828 101777-832
4-Fluorotoluene ≥97%
4-Fluorotoluene

TCA listed: T
Hazard: Flammable

Formula: C₇H₇F
MW: 110.13 g/mol
Boiling Pt: 116°C
Melting Pt: -56°C
Density: 1
MDL Number: MFCD00000358
CAS Number: 352-32-9
UN: 2388
ADR: 3,II

Order Now

Specification Test Results

Boiling point 116°
Density 1
ND20 1.468

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR