Perfluorooctanoic anhydride ≥97%

Supplier: Matrix Scientific

004854-1G 004854-5G 004854-25G
101777-790EA 138.46 USD
101777-790 101777-792 101777-794
Perfluorooctanoic anhydride ≥97%
Perfluorooctanoic anhydride

TCA listed: T
Hazard: Corrosive

Formula: C₁₆F₃₀O₃
Boiling Pt: 100 °C (15 mmHg)
MDL Number: MFCD00039243
CAS Number: 33496-48-9

Order Now

Specification Test Results

Boiling point 100°/15mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR