Hexafluoroacetone trihydrate ≥98%
Supplier: Matrix Scientific
Synonyms:
Perfluoroacetone trihydrate
004821-100G
004821-25G
101777-772EA
104.62
USD
101777-772
101777-770
Hexafluoroacetone trihydrate ≥98%
Hexafluoroacetone trihydrate
Hazard: Toxic/Irritant-Harmful
Formula:
C₃F₆O·3H₂O MW: 220.07 g/mol Melting Pt: 18…21 °C Density: 1.579 |
MDL Number:
MFCD00149061 CAS Number: 34202-69-2 UN: 2552 ADR: 6.1 II |
Specification Test Results
Melting point | 18-21° |
Density | 1.579 |
ND20 | 1.319 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...