4-Fluoro-o-xylene ≥99%
Supplier: Matrix Scientific
Synonyms:
1-Fluoro-3,4-dimethylbenzene, 4-Fluoro-1,2-dimethylbenzene
004815-5G
004815-25G
101777-760EA
30.77
USD
101777-760
101777-762
4-Fluoro-o-xylene ≥99%
4-Fluoro-o-xylene
Hazard: Flammable
Formula:
C₈H₉F MW: 124.16 g/mol Boiling Pt: 50°C/20mm |
MDL Number:
MFCD00039217 CAS Number: 452-64-2 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 50°/20mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...