4-Nitro-α,α,α-trifluorotoluene ≥98%
Supplier: Matrix Scientific
004749-5G
004749-25G
101777-732EA
120
USD
101777-732
101777-734
4-Nitro-α,α,α-trifluorotoluene ≥98%
4-Nitro-α,α,α-trifluorotoluene
Hazard: Toxic
Formula:
C₇H₄F₃NO₂ Boiling Pt: 81…82 °C (10 mmHg) Melting Pt: 40…41 °C |
MDL Number:
MFCD00007358 CAS Number: 402-54-0 UN: 3431 ADR: 6.1,II |
Specification Test Results
Boiling point | 81-82°/10m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...