4-Bromo-α,α,α-trifluorotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Bromobenzotrifluoride, 1-Bromo-4-(trifluoromethyl)benzene

004745-100G 004745-500G
101777-724EA 76.92 USD
101777-724 101777-726
4-Bromo-α,α,α-trifluorotoluene ≥98%
4-Bromo-α,α,α-trifluorotoluene

TCA listed: T
Hazard: Flammable

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 154…155 °C
Density: 1.607
MDL Number: MFCD00000398
CAS Number: 402-43-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 154-155°
Density 1.607
ND20 1.473

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR