2-Bromobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromobenzene-1-sulphonyl chloride

004725-25G 004725-100G
101777-708EA 100 USD
101777-708 101777-710
2-Bromobenzenesulfonyl chloride ≥98%
2-Bromobenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Melting Pt: 51-52°C
MDL Number: MFCD00051971
CAS Number: 2905-25-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 51-52°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR