4-Bromocinnamic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(4-Bromophenyl)acrylic acid

004661-25G 004661-100G
101777-686EA 26.15 USD
101777-686 101777-688
4-Bromocinnamic acid ≥98%
4-Bromocinnamic acid

Hazard: Irritant

Formula: C₉H₇BrO₂
MW: 227.06 g/mol
Melting Pt: 262…264 °C
MDL Number: MFCD00004394
CAS Number: 1200-07-3

Order Now

Specification Test Results

Melting point 262-264°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR