Thiophen-2-acetonitrile ≥96%
Supplier: Matrix Scientific
Synonyms:
Thiophene-2-acetonitrile, 2-Thiopheneacetonitrile, 2-(Thiophen-2-yl)acetonitrile
004621-25G
004621-100G
101777-648EA
20
USD
101777-648
101777-650
Thiophen-2-acetonitrile ≥96%
Thiophen-2-acetonitrile
TCA listed: T
Hazard: Toxic
Formula:
C₆H₅NS MW: 123.18 g/mol Boiling Pt: 115-120°C/22mm Density: 1.157 |
MDL Number:
MFCD00005453 CAS Number: 20893-30-5 UN: 3276 ADR: 6.1,III |
Specification Test Results
Boiling point | 115-120°/2 |
Density | 1.157 |
ND20 | 1.542 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...