1-Bromononane ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Nonyl bromide

004606-25G 004606-100G
101777-644EA 26.15 USD
101777-644 101777-646
1-Bromononane ≥98%
1-Bromononane

TCA listed: T
Hazard: Irritant

Formula: BrH₂C(CH₂)₇CH₃
Boiling Pt: 201°C
Density: 1.084
Flash Pt: 90°C
MDL Number: MFCD00000278
CAS Number: 693-58-3

Order Now

Specification Test Results

Boiling point 201°
Density 1.084
ND20 1.454


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR