3,4-Dibromobenzenesulfonyl chloride
Supplier: Matrix Scientific
Synonyms:
3,4-Dibromobenzene-1-sulphonyl chloride
004568-1G
004568-5G
101777-630EA
53.85
USD
101777-630
101777-632
3,4-Dibromobenzenesulfonyl chloride
3,4-Dibromobenzenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive/Lachrymator
Formula:
C₆H₃Br₂ClO₂S MW: 334.42 g/mol Melting Pt: 34 °C |
MDL Number:
MFCD00060709 CAS Number: 81903-80-2 |
Specification Test Results
Melting point | 34° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...