2,4-Dibromoaniline ≥98%

Supplier: Matrix Scientific

004563-25G 004563-100G
101777-622EA 29.23 USD
101777-622 101777-624
2,4-Dibromoaniline ≥98%
2,4-Dibromoaniline

TCA listed: T
Hazard: Toxic

Formula: C₆H₅Br₂N
Melting Pt: 78…80 °C
Density: 2.297
MDL Number: MFCD00007633
CAS Number: 615-57-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 78-80°
Density 2.297


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR