1,1-Dibromo-3,3,3-trifluoroacetone ≥95%

Supplier: Matrix Scientific

Synonyms: 3,3-Dibromo-1,1,1-trifluoroacetone, 3,3-Dibromo-1,1,1-trifluoro-2-propanone, 3,3-Dibromo-1,1,1-trifluoropropan-2-one

004542-100G 004542-25G
101777-616EA 269.23 USD
101777-616 101777-614
1,1-Dibromo-3,3,3-trifluoroacetone ≥95%
1,1-Dibromo-3,3,3-trifluoroacetone

Hazard: Toxic/Refrigerate/Lachrymator

Formula: Br₂HCC(O)CF₃
MW: 269.84 g/mol
Boiling Pt: 111-113°C
MDL Number: MFCD00041362
CAS Number: 431-67-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 111-113°
ND20 1.4297

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR