Triethylammonium dihydrogentrifluoride ≥98%

Supplier: Matrix Scientific

Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride

004509-25G 004509-100G
101777-606EA 24.62 USD
101777-606 101777-608
Triethylammonium dihydrogentrifluoride ≥98%
Triethylammonium dihydrogentrifluoride

TCA listed: T
Hazard: Toxic/Corrosive/Stored under Argon/Hygroscopic/Refrigerate

Formula: C₆H₁₈F₃N
MW: 161.21 g/mol
Boiling Pt: 70°C/15mm
Melting Pt: -27°C
Density: 1.007/20°C
MDL Number: MFCD00043294
CAS Number: 73602-61-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 70°/15mm
Density 0.989
ND20 1.3915

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR