(Trifluoromethyl)trimethylsilane ≥96%

Supplier: Matrix Scientific

Synonyms: trimethyl(trifluoromethyl)silane

004503-25G 004503-100G
101777-602EA 69.23 USD
101777-602 101777-604
(Trifluoromethyl)trimethylsilane ≥96%
(Trifluoromethyl)trimethylsilane

Hazard: Flammable/Refrigerate

Formula: C₄H₉F₃Si
MW: 142.2 g/mol
Boiling Pt: 54-55°C
Density: 0.962
Flash Pt: -10°C
MDL Number: MFCD00145454
CAS Number: 81290-20-2

Order Now

Specification Test Results

Boiling point 45°
Density 0.962
ND20 1.33

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR